Skip to Content
MilliporeSigma

SML4161

Enrofloxacin hydrochloride

≥98% (HPLC), powder, Fluoroquinolone antibiotic

Synonym(s):

1-Cyclopropyl-7-(4-ethyl-1-piperazinyl)-6-fluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid hydrochloride, Enrofloxacin HCl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H22FN3O3.HCl
CAS Number:
Molecular Weight:
395.86
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

OC(C1=CN(C2CC2)C3=CC(N4CCN(CC)CC4)=C(F)C=C3C1=O)=O.Cl

Application

Enrofloxacin hydrochloride may be used in studies assessing the presence of enrofloxacin in water sources, contributing to environmental safety and pollution control. It may also be used in analytical methods for detecting enrofloxacin residues in food products, ensuring compliance with safety regulations.

Biochem/physiol Actions

Enrofloxacin hydrochloride is a fluoroquinolone antibiotic with broad-spectrum activity, which kills bacteria by blocking DNA gyrase and type IV topoisomerase interaction with DNA. This antibiotic belongs to the fluoroquinolone group, specifically the 6-fluoro-7-piperazinyl-4-quinolones. Enrofloxacin hydrochloride is effective against both Gram-negative and Gram-positive bacteria. It is commonly used in veterinary medicine. Enrofloxacin HCl exhibits better water solubility than enrofloxacin.


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library