Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C14H11ClN2O
CAS Number:
Molecular Weight:
258.70
NACRES:
NA.21
Assay:
≥95% (HPLC)
Form:
powder
Quality Segment
assay
≥95% (HPLC)
form
powder
color
, Faint green to light yellow brown
solubility
DMSO: soluble mg/mL, clear
storage temp.
-10 to -25°C
SMILES string
ClC(C=CC1=C2N)=CC1=NC3=C2C=C(OC)C=C3
Application
9-Amino-6-chloro-2-methoxyacridine (ACMA) may be used in imaging and labeling applications in biological research, particularly for studying cellular processes and interactions.
Biochem/physiol Actions
9-Amino-6-chloro-2-methoxyacridine (ACMA) is a versatile fluorescent molecule that exhibits pH-sensitive fluorescence, with high intensity at physiological pH and quenching upon protonation, coupled with decreased membrane permeability. 9-ACMA functions as a DNA intercalator, preferentially binding to poly(dA-dT) sequences over poly(dG-dC), with excitation/emission maxima around 410/480 nm, compatible with various UV light sources. ACMA interacts with energized membranes and undergoes fluorescence quenching in response to transmembrane pH gradients. It is extensively employed to monitor cation and anion movement across membranes and to study the proton-pumping activity of various membrane-bound ATPases. ACMA has the capability to induce frameshift mutations as well as transversion and transition mutations. Additionally, it can cause base-pair substitutions in the presence of S9 mix.
signalword
Warning
hcodes
Hazard Classifications
Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

