Skip to Content
MilliporeSigma

SML4163

BODIPY 558/568 C12, Red

≥98% (HPLC), (Powder, film, or oil), fluorescent probe

Synonym(s):

3-(11-Carboxyundecyl)-5,5-difluoro-7-(thiophen-2-yl)-5H-6l4-dipyrrolo[1,2-c:2′,1′-f][1,3,2]diazaborinin-5-uide, (T-4)-difluoro[5-[[5-(2-thienyl)-2H-pyrrol-2-ylidene-κN]methyl]-1H-pyrrole-2-dodecanoato(2-)-κN1]-borate(1-) monohydrogen, Hydrogen (T-4)-difluoro[5-[[5-(2-thienyl)-2H-pyrrol-2-ylidene-κN]methyl]-1H-pyrrole-2-dodecanoato(2-)-κN1]borate(1-), BDP 558/568 C12

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H31BF2N2O2S
CAS Number:
Molecular Weight:
472.40
NACRES:
NA.21
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
(Powder, film, or oil)
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

(Powder, film, or oil)

storage condition

desiccated

color

red to black

storage temp.

-10 to -25°C

SMILES string

F[B-]1([N+]2=C(C3=CC=CS3)C=CC2=CC4=CC=C(CCCCCCCCCCCC(O)=O)N14)F

General description

BODIPY 558/568 C12, also known as BODIPY-red C12, is an orange-red fluorescent saturated fatty acid analog structurally resembling an 18-carbon fatty acid.

Application

BODIPY 558/568 C12, Red is suitable for use in:
  • investigating lipid dynamics, such as the formation, growth, and breakdown of lipid droplets
  • fluorescence microscopy techniques
  • cell labeling following molecules throughout cellular processes
  • studying protein–protein interactions

Biochem/physiol Actions

BODIPY 558/568 C12 monitors lipid flux/fatty acid trafficking in live cells and fixed cells. In cells, red-C12 probe primarily localizes in neutral lipids within lipid droplets (LDs) and to the endoplasmic reticulum (ER), tracing lipid accumulation in these structures as well as in mitochondria and peroxisomes. BDP 558/568 C12 reveals fatty acid transfer from lipid droplet triacylglycerides (TAGs) to mitochondria and demonstrates how these fatty acids are esterified and stored in lipid droplets within the cytotrophoblast layer. Red C12 features a sharp fluorescence peak and high quantum yield, maintaining stable fluorescence across various physiological conditions and resisting changes in environmental polarity and pH. Excites at ~558 nm and emits at ~568 nm. BODIPY = boron dipyrromethene.

Disclaimer

Hygroscopic Store with dessicant


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library