Skip to Content
MilliporeSigma

SML4167

HMSiR680-Me

≥90% (HPLC)

Synonym(s):

N,1′,11′,13′,13′-Pentamethyl-2′,3′,4′,8′,9′,10′,11′,13′-octahydro-1′H,3H-spiro[isobenzofuran-1,6′-silino[3,2-g:5,6-g′]diquinoline]-5-carboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C32H37N3O2Si
Molecular Weight:
523.74
UNSPSC Code:
12352200
Assay:
≥90% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥90% (HPLC)

form

powder

color

dark blue to black

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

HMSiR680-Me is a cell-permeable silicon rhodamine (SiR) far-red fluorophore that selectively illuminates acidic organelles, such as lysosomes, in live cells, owing to its unique pH-dependent fluorescence. It is ideal for complex imaging experiments, offering high photostability and minimal impact on cell health or organelle motility. HMSiR680-Me enables multicolor imaging alongside other dyes, with protocols for crosstalk calculation and spectral unmixing to ensure clear and distinct signals. Its utility extends to various applications, including overnight imaging and super-resolution microscopy, making HMSiR680-Me a valuable tool for studying cellular processes and diseases related to lysosomal function. Ex: ~ 680 nm & Em: ~ 695 nm; Detection window ~ 690-830 nm; pKcyclic = 6.2.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library