Skip to Content
MilliporeSigma

SML4174

Avatrombopag

≥98% (HPLC), powder, Thrombopoietin receptor (TPOR) agonist

Synonym(s):

1-[3-Chloro-5-[[[4-(4-chloro-2thienyl)-5-(4-cyclohexyl-1-piperazinyl)-2-thiazolyl]amino]carbonyl]-2-pyridinyl]-4-piperidinecarboxylic acid, 1-(3-Chloro-5-{[4-(4-chloro-2-thienyl)-5-(4-cyclohexylpiperazin-1-yl)-1,3-thiazol-2-yl]carbamoyl}-2-pyridyl)piperidine-4-carboxylic acid, AKR 501, AKR-501, AKR501, E 5501, E-5501, E5501, YM 477, YM-477, YM477

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C29H34Cl2N6O3S2
CAS Number:
Molecular Weight:
649.65
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

ClC1=CC(C(NC2=NC(C3=CC(Cl)=CS3)=C(N4CCN(C5CCCCC5)CC4)S2)=O)=CN=C1N6CCC(C(O)=O)CC6

Application

Avatrombopag may be used in laboratory studies to explore the mechanisms of thrombopoiesis and the role of thrombopoietin receptor agonists in platelet production.

Biochem/physiol Actions

Avatrombopag (E5501; YM477; AKR 501) is an orally active, potent, and human and chimpanzee species-specific thrombopoietin (TPO) receptor (TPOR) agonist that induces cellular STAT3/5 phosphorylation and proliferation of hTPOR-expressing Ba/F3 cells (proliferation EC50 = 3.3 nM). Avatrombopag promotes megakaryocyte differentiation from human cord blood (CB) CD34+ cells (EC50 = 25 nM) and increases the number of human platelets in NOD/SCID mice transplanted with human fetal liver CD34(+) cells in vivo (0.3, 1, 3 mg/kg/d p.o.). This small molecule agonist of the TPO‐RA enhances platelet production by activating the receptor and mimicking the biological effects of thrombopoietin (TPO). Its distinctive chemical structure allows for selective binding to the transmembrane domain of the TPO receptor (TPO-R), preventing interference with the binding of endogenous TPO. Avatrombopag activates crucial intracellular pathways, such as the phosphorylation of STAT3, STAT5, and ERK (MAPK), which promote the differentiation and proliferation of megakaryocytes, a vital process for platelet production.
Orally active, potent and human and chimpanzee species-specific thrombopoietin receptor (TPO) agonist in vitro and in vivo.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library