Skip to Content
MilliporeSigma

SML4178

Maleimide-PEG2-biotin

≥98% (HPLC), powder, biotinylation reagent

Synonym(s):

(3aS,4S,6aR)-N-[2-[2-[2-[[3-(2,5-Dihydro-2,5-dioxo-1H-pyrrol-1-yl)-1-oxopropyl]amino]ethoxy]ethoxy]ethyl]hexahydro-2-oxo-1H-thieno[3,4-d]imidazole-4-pentanamide, Biotin-PEG2-Mal, Biotin-PEO-maleimide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H35N5O7S
CAS Number:
Molecular Weight:
525.62
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=C(N1CCC(NCCOCCOCCNC(CCCC[C@H]2[C@@]3([C@]([H])(CS2)NC(N3)=O)[H])=O)=O)C=CC1=O

General description

Maleimide-PEG2-biotin, a maleimide-conjugated biotin molecule with a medium-length spacer arm (PEG)2, is membrane-impermeable and exhibits high affinity for free thiol groups on cysteines.

Application

Maleimide-PEG2-biotin is suitable for use in labeling and purifying biomolecules with high specificity. It may be used in mass spectrometry to detect free sulfhydryl residues in intact proteins.

Biochem/physiol Actions

Biotin-PEG2-Mal, a biotin maleimide linker, forms stable thioether bonds by covalently labeling cysteine sulfhydryl groups on antibodies or proteins and can be detected via streptavidin-HRP in Western blot assays following pull-down. Maleimide labels react with sulfhydryl (–SH) groups at acidic pH, minimizing the risk of disulfide bond scrambling during labeling that can happen at higher pH levels.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library