Skip to Content
MilliporeSigma

SML4182

Cypate

≥90% (HPLC)

Synonym(s):

3-​(2-​carboxyethyl)​-​2-​[7-​[3-​(2-​Carboxyethyl)​-​1,​3-​dihydro-​1,​1-​dimethyl-​2H-​benz[e]​indol-​2-​ylidene]​-​1,​3,​5-​heptatrien-​1-​yl]​-​1,​1-​dimethyl-​1H-​benz[e]​indolium chloride, Cy-2-glu

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C41H41N2O4·Cl
CAS Number:
Molecular Weight:
661.23
UNSPSC Code:
12352200
Assay:
≥90% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥90% (HPLC)

form

powder

storage condition

desiccated

color

green to black

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Cypate is a cell penetrant near-infrared fluorescence probe. It appears that cypate uses a non-glucose-utilizing pathway for transport such as fatty acid transport pathway. Cypate is suitable for conjugation with different agents, tracers and probes for optical imaging of tumors and other physiological processes.

Disclaimer

Hygroscopic, store with desiccant


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library