Skip to Content
MilliporeSigma

SML4185

Gadoxetate disodium

≥95% (Gadolinium, elemental analysis), powder, contrast agent

Synonym(s):

Gadoxetic acid disodium salt, Gd-EOB-DTPA disodium, Gadolinium-ethoxybenzyl-diethylenetriamine pentaacetic acid disodium, ZK 139834, N-[2S-2-[Bis(carboxymethyl)amino]-3-(4-ethoxyphenyl)propyl]-N-[2-[bis(carboxymethyl)amino]ethyl]-glycine gadolinium complex disodium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H28GdN3O11· 2Na
CAS Number:
Molecular Weight:
725.71
NACRES:
NA.21
Assay:
≥95% (Gadolinium, elemental analysis)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (Gadolinium, elemental analysis)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

[O-]C(CN(CCN(C[C@H](CC1=CC=C(C=C1)OCC)N(CC([O-])=O)CC([O-])=O)CC([O-])=O)CC([O-])=O)=O.[Gd+3].[Na+].[Na+]

General description

Gadoxetate disodium is an amphipathic derivative of gadolinium diethylenetriaminepentaacetic acid (Gd-DTPA). It is highly water soluble, hepatocyte-specific paramagnetic gadolinium-based MRI contrast agent. Structurally, it is an ionic, linear gadolinium-based agent with a lipophilic ethoxybenzyl (EOB) moiety covalently attached to gadolinium diethylenetriaminepentaacetic acid (Gd-DTPA).

Application

Gadoxetate disodium may be used in liver and hepatobiliary imaging by enhancing the visualization of liver lesions, including tumors and metastases, and helping in assessing hepatobiliary disorders by providing detailed images of bile ducts and liver function.

Biochem/physiol Actions

Dual-excretion MRI contrast agent for liver-specific imaging, enhancing tumor detection, hepatic and biliary function assessment.

It is designed for enhanced imaging of liver structures and functions, with a unique dual excretion profile of 50% renal and 50% hepatic elimination. Its unique structure enables uptake by hepatocytes through the organic anion transporting polypeptide (OATP) and subsequent excretion into bile via multidrug resistance-associated proteins (MRPs). Gadoxetate uniquely distributes into vascular, extracellular, and hepatocellular spaces, allowing dual-function imaging to assess both liver function and biliary drainage while providing T1 relaxivity of 6.9 l mmol−1 s−1 at 1.5 T (in plasma). Clinically, this agent (intravenous bolus injection of 0.025 mmol/kg (0.1 mL/kg) body weight) is critical for detecting and characterizing liver tumors, evaluating hepatic function, and visualizing biliary system abnormalities, offering stable and targeted imaging with reduced gadolinium toxicity risk.

Disclaimer

Hygroscopic Store with desiccant


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library