Skip to Content
MilliporeSigma

SML4192

DNA2 Inhibitor C5

≥95% (HPLC), powder, DNA2 helicase/nuclease inhibitor

Synonym(s):

4-Hydroxy-8-nitroquinoline-3-carboxylic acid, 4-Hydroxy-8-nitro-3-quinolinecarboxylic acid, DNA2i-C5, NSC 15765

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H6N2O5
CAS Number:
Molecular Weight:
234.17
MDL number:
NACRES:
NA.21
Assay:
≥95% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=C(C1=CN=C2C(C=CC=C2[N+]([O-])=O)=C1O)O

Application

DNA2 Inhibitor C5 may be used to evaluate the on-target effects on DNA2 in cancer cells.

Biochem/physiol Actions

DNA2 Inhibitor C5 suppresses nuclease/helicase activities, impairs DNA repair/replication, causes cell cycle arrest, and shows synergistic cancer cell killing with DNA-damaging agents and PARP/ATR inhibitors.

The DNA2 inhibitor C5 (DNA2i-C5) binds to a pocket near the DNA binding site in the helicase domain of DNA2 with an IC50 of 20-30 μM for inhibiting nuclease activity and DNA binding. C5 effectively suppresses both the nuclease and helicase activities of DNA2, impairing DNA end resection, homologous recombination, single-strand annealing repair, and replication fork restart after stalling. In cells, C5 mimics genetic knockout of DNA2, causing cell cycle arrest in late S/G2 phase, reducing mitotic entry, and leading to chromosomal segregation defects, particularly affecting centromeric DNA replication and integrity. Importantly, C5 shows synergistic killing of various cancer cell types when combined with DNA damaging agents like camptothecin or inhibitors of PARP and ATR, highlighting its potential as a cancer therapeutic strategy.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library