Skip to Content
MilliporeSigma

SML4199

5-Hydroxymethylcytosine

≥95% (NMR)

Synonym(s):

4-Amino-2-hydroxy-5-(hydroxymethyl)pyrimidine, 5-(Hydroxymethyl)cytosine; 5-(Oxymethyl)cytosine, 6-Amino-5-(hydroxymethyl)-2(1H)-pyrimidinone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H7N3O2
CAS Number:
Molecular Weight:
141.13
UNSPSC Code:
12352200
Assay:
≥95% (NMR)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (NMR)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

5-Hydroxymethylcytosine (5hmC), a relatively stable cytosine modification, is an epigenetic mark that regulate vertebrate gene expression. 5-Hydroxymethylcytosine is enriched at enhancers in mouse and human embryonic stem cells (mESCs and hESCs).


Still not finding the right product?

Explore all of our products under 5-Hydroxymethylcytosine


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library