Skip to Content
MilliporeSigma

SML4205

I-BET787

≥98% (HPLC), powder, pan-BET bromodomain inhibitor

Synonym(s):

5-[4-Chloro-1-[(tetrahydro-2H-pyran-4-yl)methyl]-1H-imidazol-2-yl]-1,3-dimethyl-2(1H)-pyridinone, 5-(4-Chloro-1-((tetrahydro-2H-pyran-4-yl)methyl)-1H-imidazole-2-yl)-1,3-dimethylpyridin-2(1H)-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H20ClN3O2
CAS Number:
Molecular Weight:
321.80
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Orally active, potent and selective pan-BET family bromodomain (BrD) inhibitor. I-BET787 is an orally active, potent and selective pan-BET bromodomain (BrD) inhibitor (BRD2/3/4/T pIC50 =6.9/7.1/7.1/6.3/BrD1 5.7/6.2/5.9/5.1/BrD2) that demonstrates efficacy in an inflammation model in mice in vivo (5 & 15 mg/kg b.i.d. p.o. for 14 days)by suppressing KLH-TNP (100 μg/kg via i.p. 1h post first I-BET787 oral dosing)-induced T-cell activation.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1B

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library