Skip to Content
MilliporeSigma

SML4206

DMXAA sodium salt

≥98% (HPLC)

Synonym(s):

5,6-Dimethyl-9-oxo-9H-xanthene-4-acetic acid sodium salt; sodium 2-(5,6-Dimethyl-9-oxo-9H-xanthen-4-yl)acetate, 2-(5,6-Dimethyl-9-oxo-9H-xanthen-4-yl)acetic acid sodium salt; 5,6-Dimethylxanthenone-4-acetic acid sodium salt, Vadimezan sodium salt, ASA404 sodium salt, NSC 640488 sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H13NaO4
CAS Number:
Molecular Weight:
304.27
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

A water-soluble sodium salt form of DMXAA. DMXAA directly binds to murine STING (stimulator of interferon genes) but does not affect human STING. DMXAA disrupts tumor vasculature by inducing endothelial cell apoptosis, causing tumor hemorrhagic necrosis and ischemia, releasing serotonin, stimulating the innate immune system, producing inflammatory cytokines (e.g., TNF, IL-6, GCSF, KC, IP-10, MCP-1), activating NF-κB and p38 MAPK, producing nitric oxide, and reducing tumor energetics and membrane turnover.

Disclaimer

Hygroscopic Store with desiccant


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library