Skip to Content
MilliporeSigma

SML4209

BHB-Phe

≥98% (HPLC), powder, BHB secondary metabolite

Synonym(s):

N-[(3R)-3-Hydroxy-1-oxobutyl]-L-Phenylalanine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H17NO4
CAS Number:
Molecular Weight:
251.28
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

C[C@@H](O)CC(N[C@@H](CC1=CC=CC=C1)C(O)=O)=O

General description

BHB-Phe is a β-Hydroxybutyrate (BHB) secondary metabolite present in human and mouse plasma. It is a ketosis-inducible structural and functional congener of lactate-phenylalanine.

Application

BHB-Phe may be used for examining its effects on metabolic pathways during states of ketosis and studying its interactions with the carnosine dipeptidase 2 (CNDP2) enzyme.

Biochem/physiol Actions

BHB-Phe exhibits anti-obesity activities hence reducing food intake and body weight in diet-induced obese (DIO) mice. Apparently BHB-Phe activates hypothalamic and brainstem neurons and suppresses feeding. BHB-Phe issynthesized in the body by catalytic condensation of BHB and Phenylalanine by carnosine dipeptidase 2 (CNDP2).


pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library