Skip to Content
MilliporeSigma

SML4211

Fluorosamine

≥98% (HPLC), (liquid to crytals), CSPG synthesis inhibitor

Synonym(s):

(3R,4R,5S,6R)-3-Acetamido-6-(acetoxymethyl)-5-fluorotetrahydro-2H-pyran-2,4-diyl diacetate, 2-Acetamido-4-fluoro-1,3,6-tri-o-acetyl-2,4-dideoxy-d-glucopyranose, peracetylated 4-F-GlcNAc, 2-Acetamido-1,3,6-tri-O-acetyl-2,4-dideoxy-4-fluoro-D-glucopyranose, Ac-4-F-GlcNAc

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H20FNO8
CAS Number:
Molecular Weight:
349.31
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
(liquid to crytals)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

(liquid to crytals)

color

white to yellow

storage temp.

2-8°C

General description

Fluorosamine is characterized by its fluorinated glucosamine structure, specifically modified at the 4-position with a fluorine atom. The presence of the fluorine atom is crucial for its inhibitory action on CSPG biosynthesis.

Application

Fluorosamine may be used in in vitro and in vivo models to study the mechanisms underlying remyelination and the role of extracellular matrix components in neural repair.

Biochem/physiol Actions

Fluorosamine (per-O-Acetylated-4-F-N-acetylglucosamine) is a cell-permeable inhibitor of chondroitin sulfate glycosaminoglycan (CS-GAG) chain assembly, reducing chondroitin sulfate proteoglycan (CSPG) activity and perineuronal nets (PNNs) in the arcuate nucleus (ARC). In vitro, at 10 µg/ml, Ac-4-F-GlcNAc significantly reduces neurofibrosis by preventing CS-GAG synthesis and elongation. In vivo, Fluorosamine administered intranasally (1 mg and 5 mg) or intracerebroventricularly (100 µg and 250 µg) depletes GanNAc, improves glycemic control, weight loss, and energy expenditure by restoring ARC insulin sensitivity in obese and diabetic mice. Additionally, it accelerates remyelination and exhibits immunomodulatory properties, reducing T-cell proliferation in CNS models.


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library