Skip to Content
MilliporeSigma

SML4212

JQKD82 dihydrochloride

≥98% (HPLC), powder, Histone Demethylase Inhibitor

Synonym(s):

2,4-Bis(1-methylethoxy)phenyl 2-[[[2-[[2-(dimethylamino)ethyl]ethylamino]-2-oxoethyl]amino]methyl]-4-pyridinecarboxylate dihydrochloride, JQKD 82 dihydrochloride, JQKD-82 dihydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C27H40N4O5·2HCl
CAS Number:
Molecular Weight:
573.55
NACRES:
NA.21
UNSPSC Code:
12352203
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=C(C1=CC(CNCC(N(CCN(C)C)CC)=O)=NC=C1)OC2=CC=C(C=C2OC(C)C)OC(C)C.Cl.Cl

General description

JQKD82 dihydrochloride is histone H3K4 demethylase inhibitor is a produg of KDM5-C49 phenol ester prodrug that selectively upregulates H3K4me3 and inhibits KDM5-dependent cancer growth.
(KDOAM-21; KDM5A/B/C/D Ki = 2/1/6.1/3.4 nM vs. KDM4C/6B/3A/2A Ki =0.51/4.55/2.59/4.4 μM; KDM5A/B/C/D IC50 = 1.1/0.8/3.2/2.7 μM by FDH assay with[αKG] = 1 mM & [E]/[S] = 0.5/15 μM; KDM5A/B/C IC50 = 25/30/59 nM by alphaLISA with [αKG] = 25 μM & [E]/[S] = 10/100 nM). JQKD82 selectively upregulates cellular histone H3K4me3 with a higher efficacy than KDM5-C49 orKDM5-C70 (3 μM for 24 h; MM.1S cells), and inhibits myeloma growth in cultures (MM.1S IC50 = 0.42 μM vs IC50 3.1 μM/KDM5-C70, >10 μM/KDM5-C49) and in vivo (50 mg/kg b.i.d. i.p. to mice with MOLP-8 xenograft).

Application

JQKD82 dihydrochloride may be used:
  • to study its effects on lysine demethylase 5A (KDM5A) function in multiple myeloma research
  • as a potential agent in HIV-1 research, where it was evaluated for its ability to reactivate latent HIV-1 inT cells
  • to study its effects on cell cycle arrest and apoptosis induction

Biochem/physiol Actions

A selective and cell-permeable lysine demethylase 5(KDM5) inhibitor.

Disclaimer

Hygroscopic


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library