Skip to Content
MilliporeSigma

SML4214

BODIPY-TMR-X NHS ester

≥95% (HPLC)

Synonym(s):

2,5-Dioxopyrrolidin-1-yl 6-(3-(5,5-difluoro-7-(4-methoxyphenyl)-1,3-dimethyl-5H-5l4,6l4-dipyrrolo[1,2-c:2′,1′-f][1,3,2]diazaborinin-2-yl)propanamido)hexanoate, (T-4)-[N-[6-[(2,5-Dioxo-1-pyrrolidinyl)oxy]-6-oxohexyl]-5-[[5-(4-methoxyphenyl)-2H-pyrrol-2-ylidene-κN]methyl]-2,4-dimethyl-1H-pyrrole-3-propanamidato-κN1]difluoroboron, BTMR-X NHS, BDY-TMR-X, BODIPY-TMR-X NHS ester, BODIPY TMR-X succinimidyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C31H35BF2N4O6
CAS Number:
Molecular Weight:
608.44
NACRES:
NA.21
UNSPSC Code:
12352200
Assay:
≥95% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

color

red to dark red-brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=C(ON1C(CCC1=O)=O)CCCCCNC(CCC2=C(C3=CC4=CC(C5=CC=C(C=C5)OC)=CN4B([N]3=C2C)(F)F)C)=O

General description

BODIPY-TMR-X NHS ester is a hydrophobic, orange fluorophore with seven-atomaminohexanoyl spacer for amine-bioconjugation.

Application

BODIPY-TMR-X NHS ester may be used as a fluorescent labeling reagent to enable visualization and tracking of molecular interactions in biological systems and imaging workflows.

Biochem/physiol Actions

The BODIPY-TMR-X NHS ester is a versatile and stable tool forbioconjugation to amine-containing molecules, providing enhanced fluorescencecharacteristics and reduced interference with biomolecules, making it ideal foradvanced microscopy techniques due to its brightness and narrow emission. This bright orange fluorophore exhibits spectral properties akin to tetramethylrhodamine (TMR, TRITC), boasting high extinction coefficients and quantum yields, while maintaining relative stability across varying solvent polarities and pH levels, with excitation and emission maxima at approximately 544 nm and 570 nm, respectively. The hydrophobic nature of BODIPY-TMR-X NHS makes it particularly effective for labeling lipophilic structures such as lipids and cell membranes. Furthermore, BDP-TMR-X’s narrow emission bandwidths, prolonged excited-state lifetime, and substantial two-photon cross-section render it highly suitable for advanced applications including fluorescence polarization assays and multiphoton microscopy.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library