Skip to Content
MilliporeSigma

SML4219

GAT2711

≥98% (HPLC)

Synonym(s):

α9 nAChR Agonist GAT2711, 1-Ethyl-1-methyl-4-(4-(6-methylpicolinamido)phenyl)piperazin-1-ium iodide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H27IN4O
CAS Number:
Molecular Weight:
466.36
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

GAT2711 is a potent, selective full agonist of α9 nicotinic acetylcholine receptors (nAChRs), exhibiting an EC50 of 230 nM and 340-fold selectivity over α7 nAChRs. Reversible binding at the α9(+)α9(-) interface is supported by molecular docking and electrophysiology studies, which show concentration-dependent inward currents in α9 nAChR-expressing Xenopus oocytes. GAT2771 demonstrates anti-inflammatory potential by inhibiting ATP-induced IL-1β release in THP-1 cells (IC50 = 0.5 μM) and human PBMCs (effective at 100 fM). In a complete Freund́s adjuvant (CFA)-induced inflammatory pain model, intraperitoneal administration (2 and 10 mg/kg) in mice, including α7 nAChR knockouts, produced potent, long-lasting (up to 72 hours) analgesic effects, suggesting an α9 nAChR-mediated mechanism for managing inflammatory and neuropathic pain.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2 - Skin Sens. 1B

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library