Skip to Content
MilliporeSigma

SML4221

Ofloxacin hydrochloride

≥98% (HPLC), powder, fluoroquinolone antibiotic

Synonym(s):

9-Fluoro-2,3-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid hydrochloride, (±)-9-Fluoro-2,3-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid hydrochloride, Ofloxacin HCl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H20FN3O4 · xHCl
CAS Number:
Molecular Weight:
361.37 (free base basis)
MDL number:
NACRES:
NA.21
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=C(O)C1=CN2C3=C(OCC2C)C(N4CCN(C)CC4)=C(F)C=C3C1=O.[xHCl]

General description

Ofloxacinhydrochloride is a broad-spectrum fluoroquinolone antibiotic that exhibitswater solubility, whereas ofloxacin itself is only sparingly soluble in aqueoussolutions.

Application

Ofloxacin hydrochloride may be used as a standard to study theadsorption behaviour of antibiotics.

Biochem/physiol Actions

Ofloxacin kills bacteria by blocking DNA gyrase and type IV topoisomerase—critical enzymes for DNA replication and transcription—leading to cell death. Ofloxacin has 100 times higher affinity for bacterial DNA gyrase than for mammalian. It is effective against a broad array of both Gram-positive and Gram-negative bacteria, in addition to Mycoplasmas.

Disclaimer

Hygroscopic Store with desiccant


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library