Skip to Content
MilliporeSigma

SML4223

Artepillin C

≥98% (HPLC), powder, CREB-CRTC2 inhibitor

Synonym(s):

(2E)-3-[4-Hydroxy-3,5-bis(3-methyl-2-buten-1-yl)phenyl]-2-propenoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H24O3
CAS Number:
Molecular Weight:
300.39
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

General description

Artepillin C (ARC) is a prenylated derivative of p-coumaric acid. It is one of the primary phenolic compounds present in Brazilian green propolis (BGP) and Baccharis dracunculifolia. It is a p-CA derivative, featuring two prenyl groups linked to the meta-positions (C3 and C5) of the phenyl ring.

Application

Artepillin C may be used to investigate its interaction with human serum albumin (HSA) and to understand its physicochemical transport mechanism in vitro.

Biochem/physiol Actions

Phenolic component of propolis that exhibits multiple activities including anti-inflammatory, anti-tumor, antimicrobial and antioxidant activities; inhibitor of CREB-CRTC2 interaction. The anti-cancer properties of ARC are due to several mechanisms, including the induction of apoptosis, cell cycle arrest, and the inhibition of p21-activated kinase 1 (PAK1).


Still not finding the right product?

Explore all of our products under Artepillin C


Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library