Skip to Content
MilliporeSigma

SML4224

Roxadustat

≥98% (HPLC)

Synonym(s):

2-[(4-Hydroxy-1-methyl-7-phenoxyisoquinoline-3-carbonyl)amino]acetic acid, (((4-Hydroxy-1-methyl-7-phenoxyisoquinolin-3-yl)carbonyl)amino)acetic acid, N-[(4-Hydroxy-1-methyl-7-phenoxy-3-isoquinolinyl)carbonyl]glycine, ASP1517, AZD9941, FG 4592, FG-4592, FG4592

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H16N2O5
CAS Number:
Molecular Weight:
352.34
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Orally active hypoxia-inducible factor (HIF) prolyl hydroxylase (HIF-PH) PHD inhibitor with erythropoiesis-stimulatory efficacy in vivo.
Roxadustat (FG-4592) is an orally active hypoxia-inducible factor (HIF) prolyl hydroxylase (HIF-PH) PHD inhibitor that enhances the expression of hypoxia early response genes in cultures and in vivo by stabilizing HIF. Roxadustat protects PC12 from TBHP-induced apoptosis by upregulating cellular HIF-1α (62% without vs 21% with 6h 20 µM roxadustat pretreatment prior to 4h 150 µM TBHP induction). Roxadustat stimulates circulating EPO production in mice in vivo (40 or 80 mg/kg p.o.).


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library