Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C27H46O2
CAS Number:
Molecular Weight:
402.65
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear (warmed)
storage temp.
-10 to -25°C
Biochem/physiol Actions
20α-Hydroxycholesterol is a brain penetrant oxysterol present in human breast milk, human placenta and the mammalian CNS, which attenuates white matter injuries in neonatal mice. 20α-Hydroxycholesterol induces oligodendrogenesis through a sonic hedgehog Gli family zinc finger signaling activation mechanism. It increases numbers of mature oligodendrocytes, improves myelination, and rescues motor deficits.
Still not finding the right product?
Explore all of our products under 20α-Hydroxycholesterol
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.