Skip to Content
MilliporeSigma

SML4231

Neratinib

≥98% (HPLC), powder, pan-HER inhibitor

Synonym(s):

(2E)-N-[4-[[3-Chloro-4-(2-pyridinylmethoxy)phenyl]amino]-3-cyano-7-ethoxy-6-quinolinyl]-4-(dimethylamino)-2-butenamide, 4-Dimethylamino-but-2-enoic acid {4-[3-chloro-4-(pyridin-2-ylmethoxy)phenylamino]-3-cyano-7-ethoxy-quinolin-6-yl}-amide, (E)-N-[4-[3-Chloro-4-[(2-pyridinyl)methoxy]anilino]-3-cyano-7-ethoxy-6-quinolinyl]-4-(dimethylamino)-2-butenamide, (E)-N-[4-[[3-Chloro-4-((pyridin-2-yl)methoxy)phenyl]amino]-3-cyano-7-ethoxyquinolin-6-yl]-4-(dimethylamino)-2-butenamide, HKI-272, HKI 272

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C30H29ClN6O3
CAS Number:
Molecular Weight:
557.04
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CN(C)C/C=C/C(NC(C=C1C(NC2=CC(Cl)=C(C=C2)OCC3=NC=CC=C3)=C4C#N)=C(C=C1N=C4)OCC)=O

General description

Neratinib is a compound featuring a quinolone core, with a lipophilic 2-pyridinylmethyl moiety at the para-position of the aniline and a lipophilic chlorine atom at the meta-position. These structural elements enhance its activity against human epidermal growth factor receptor 2 (HER2).

Application

Neratinib may be used to study its anti-proliferative effects in gastric adenocarcinoma cells.

Biochem/physiol Actions

Irreversible pan-HER inhibitor, covalently targets EGFR/HER2/HER4, providing sustained signaling blockade and clinical benefit in HER2-positive breast cancer.

Neratinib (HKI-272), an oral, potent, irreversible pan-HER tyrosine kinase inhibitor, targets EGFR, HER2, and HER4 by covalently binding specific cysteine residues (Cys773/EGFR, Cys805/HER2) within the ATP-binding pocket. This mechanism provides prolonged target inhibition, effectively blocking HER phosphorylation (IC50 ~3-5 nM) and downstream MAPK/Akt signaling (IC50 ~2 nM). Consequently, it reduces cyclin D1/Rb phosphorylation (IC50 ~9 nM) and induces G1-S arrest and apoptosis in sensitive cell lines (e.g., BT474, SKBr3, A431) at low nM concentrations, underpinning its use against HER2-driven cancers.
Neratinib (HKI-272), an oral, potent, irreversible pan-HER tyrosine kinase inhibitor, targets EGFR, HER2, and HER4 by covalently binding specific cysteine residues (Cys773/EGFR, Cys805/HER2) within the ATP-binding pocket. This mechanism provides prolonged target inhibition, effectively blocking HER phosphorylation (IC50 ~3-5 nM) and downstream MAPK/Akt signaling (IC50 ~2 nM). Consequently, it reduces cyclin D1/Rb phosphorylation (IC50 ~9 nM) and induces G1-S arrest and apoptosis in sensitive cell lines (e.g., BT474, SKBr3, A431) at low nM concentrations, underpinning its use against HER2-driven cancers.


pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

Repr. 2

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library