Skip to Content
MilliporeSigma

SML4236

JX225

≥98% (HPLC), powder

Synonym(s):

2-(4-Bromo-3-methylphenoxy)-N-propylacetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H16BrNO2
CAS Number:
Molecular Weight:
286.16
NACRES:
NA.21
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

General description

Potent and selective high-affinity allosteric inhibitor of amino acid transporter B0AT1 (SLC6A19) (IC50 = 31nM).
JX225 is a potent and selective high-affinity allosteric inhibitor of amino acid transporter B0AT1 (SLC6A19) (IC50 = 31nM). JX225 binds to an allosteric binding site in the vestibule of the transporter, which prevents conformational change necessary for channel opening.


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library