Skip to Content
MilliporeSigma

SML4238

Rhod-2 AM

≥80% (HPLC)

Synonym(s):

9-[4-[bis[2-[(Acetyloxy)methoxy]-2-oxoethyl]amino]-3-[2-[2-[bis[2-[(acetyloxy)methoxy]-2-oxoethyl]amino]-5-methylphenoxy]ethoxy]phenyl]-3,6-bis(dimethylamino)-xanthylium bromide, [9-[4-[bis[2-(acetyloxymethoxy)-2-oxoethyl]amino]-3-[2-[2-[bis[2-(acetyloxymethoxy)-2-oxoethyl]amino]-5-methylphenoxy]ethoxy]phenyl]-6-(dimethylamino)xanthen-3-ylidene]-dimethylazanium bromide, Rhod-2/AM, Rhod 2AM, Rhod-2 AM ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C52H59N4O19·Br
CAS Number:
Molecular Weight:
1123.94
UNSPSC Code:
12352200
Assay:
≥80% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥80% (HPLC)

form

powder

storage condition

desiccated

color

red-purple to very dark purple

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Rhod 2AM-2 AM is a cell permeable acetoxymethyl ester of red fluorescent dye Rhod-2, which is used as reagent for monitoring calcium. Upon cell entry the ester accumulates in mitochondria and is hydrolyzed by cellular esterases to Rhod-2. Rhod 2AM-2 AM excitation maximum is 557 nm and emission maximum is 581 nm.

Disclaimer

Hygroscopic, store with desiccant


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library