Skip to Content
MilliporeSigma

SML4243

Veliparib

≥98% (HPLC), powder, PARP inhibitor

Synonym(s):

2-[(2R)-2-Methyl-2-pyrrolidinyl]-1H-benzimidazole-7-carboxamide, 2-[(2R)-2-Methylpyrrolidin-2-yl]-1H-1,3-benzodiazole-4-carboxamide, 2-[(2R)-2-Methylpyrrolidin-2-yl]-1H-benzimidazole-4-carboxamide, ABT-888, A 861695, ABT 888

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H16N4O
CAS Number:
Molecular Weight:
244.29
MDL number:
NACRES:
NA.21
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

-10 to -25°C

Application

Veliparib may be used in cell proliferation and colony forming assay to study its effect on O 6-methylguanine methyltransferase (MGMT) unmethylated glioblastoma.

Biochem/physiol Actions

Veliparib (ABT-888) is an orally active, potent and selective poly(ADP-ribose) polymerase (PARP) inhibitor (Ki = 5.2 nM PARP1, and 2.9 nM PARP2) that exhibits potent anti-cancer activity. Veliparib blocks DNA damage repair in cancer cells, additionally rendering them more susceptible for other anti-cancer drugs. As a potent PARP1 and PARP2 inhibitor it induces synthetic lethality in cancer cells.


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library