Skip to Content
MilliporeSigma

SML4244

GSK0660

≥98% (HPLC), powder, PPARβ/δ antagonist

Synonym(s):

3-[[[2-Methoxy-4-(phenylamino)phenyl]amino]sulfonyl]-2-thiophenecarboxylic acid methyl ester, Methyl 3-({[2-(methoxy)-4 phenyl]amino}sulfonyl)-2-thiophenecarboxylate, GSK 0660, GSK-0660

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H18N2O5S2
CAS Number:
Molecular Weight:
418.49
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Application

GSK0660 may be used in cell viability assay, real time cell index analysis, and oxyblot assays to study its neuroprotective effect on Parkinson’s disease model in vivo and in vitro.

Biochem/physiol Actions

GSK0660 is a potent and selective peroxisome proliferator-activated receptor delta (PPARdelta) and PPARbeta antagonist (IC50 = 300 nM against GW1516-induced PPARβ/δ reporter activity; IC50 = 155 nM vs >10 µM for PPARα and PPARγ by GW1516 displacement binding assays). GSK0660 effectively antagonizes PPARβ/δ agonist GW501516 (GW1516)-induced target genes expression in multiple cell types, including HuH7, rat L6 myotubes, and mouse BMDMs (1-10 nM GW1516, 100-1000 nM GSK0660).


Still not finding the right product?

Explore all of our products under GSK0660


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

No data available

flash_point_c

No data available



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library