Skip to Content
MilliporeSigma

SML4245

AT2R agonist C21

≥98% (HPLC), angiotensin II receptor agonist, powder

Synonym(s):

Buloxibutid, AT2R agonist C 21, AT2R agonist C-21, AT2R agonist compound 21, M 24, M024, Butyl [3-[4-[(1H-imidazol-1-yl)methyl]phenyl]-5-isobutylthiophen-2-yl]sulfonylcarbamate, N-Butyloxycarbonyl-3-[4-(imidazol-1-ylmethyl)phenyl]-5-isobutylthiophene-2-sulfonamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H29N3O4S2
CAS Number:
Molecular Weight:
475.62
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Application

AT2R agonist C21 may be used to study:
  • its vasorelaxant effect of C21 in mouse mesenteric arteries derived from wildtype and AT2R-knockout (AT2R-/y) mice
  • its effects on U46619-induced β-arrestin recruitment to TP-receptors and the effect of C21 on U46619-induced platelet aggregation

Biochem/physiol Actions

C21 (Buloxibutid; Compound 21; M024) is a potent and selective angiotensin II receptor (AT2R) nonpeptide agonist (Ki = 0.4 nM/AT1R vs. >10 µM/AT1R) that induces AT2R-mediated morphological changes in NG 108-15 neuroblastoma-glioma hybrid cells (100 nM for 3 days), blockable by AT2R antagonist PD 123,319 (1 µM). C21 enhances in vivo duodenal alkaline secretion in Sprague-Dawley rats (30 & 300 µg/kg/h via i.v.), and lowers the mean arterial blood pressure in anesthetized, spontaneously hypertensive rats in vivo (50 µg/kg/h via i.v.). C21 shares structural similarities with AT1-receptor antagonists irbesartan and losartan, and may show TP-receptor antagonistic properties.


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library