Skip to Content
MilliporeSigma

SML4249

EPZ020411

≥98% (HPLC)

Synonym(s):

EPZ 020411, EPZ-020411, N1,N2-Dimethyl-N1-[[3-[4-[[trans-3-[2-(tetrahydro-2H-pyran-4-yl)ethoxy]cyclobutyl]oxy]phenyl]-1H-pyrazol-4-yl]methyl]-1,2-ethanediamine (ACI)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H38N4O3
CAS Number:
Molecular Weight:
442.59
Assay:
≥98% (HPLC)
Form:
(Oil (syrup))
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

(Oil (syrup))

color

colorless to faint brown

storage temp.

-10 to -25°C

Biochem/physiol Actions

EPZ020411 is a potent and selective protein arginine methyltransferase PRMT6 inhibitor (IC50 = 10 nM; PRMT1/8 IC50 = 119/223 nM). EPZ020411 inhibits PRMT6 expression-induced H3R2 methylation in A375 cells (IC50 = 637 nM), but not PRMT1-specific methylmark (IC50 = 7.1 μM).


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library