Skip to Content
MilliporeSigma

SML4251

AM1710

≥97% (HPLC), powder, CB2 agonist

Synonym(s):

3-(1,1-Dimethylheptyl)-1-hydroxy-9-methoxy-6H-dibenzo[b,d]pyran-6-one, AM 1710

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H28O4
CAS Number:
Molecular Weight:
368.47
UNSPSC Code:
12352203
eCl@ss:
32150313
Assay:
≥97% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

General description

AM1710, a CB2 agonist, belongs to the class of cannabinoids known as cannabilactones. Cannabilactones are characterized by the presence of a carbonyl group instead of the 6,6-dimethyl group typically found in the classical tricyclic structure of cannabinoids.

Application

AM1710 may be used to study the effects of CB2 agonists on neuropathic nociception induced by the antiretroviral agent Zalcitabine, in a mouse model.

Biochem/physiol Actions

AM1710 is a highly potent and selective CB2 agonist that displays 54-fold selectivity for CB2 over CB1. AM1710 is a low potency inverse agonist/antagonist at CB1 in vitro but is not a functional CB1 antagonist in vivo. It inhibits chemotherapy-induced neuropathic pain in rodents while delaying the development of morphine tolerance and reduced naloxone-precipitated opioid withdrawal.


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library