Skip to Content
MilliporeSigma

SML4258

Spexin 1 trifluoroacetate

≥95% (HPLC), (Powder or Lyophilized powder or film), GAL2/3 agonist

Synonym(s):

H-Asn-Trp-Thr-Pro-Gln-Ala-Met-Leu-Tyr-Leu-Lys-Gly-Ala-Gln-NH₂ trifluoroacetate, L-Asparaginyl-L-tryptophyl-L-threonyl-L-prolyl-L-glutaminyl-L-alanyl-L-methionyl-L-leucyl-L-tyrosyl-L-leucyl-L-lysylglycyl-L-alanyl-L-glutamamide trifluoroacetate, Spexin trifluoroacetate, SPX trifluoroacetate, SPX1 trifluoroacetate, Neuropeptide Q trifluoroacetate, NPQ trifluoroacetate, NPQ 53-70 trifluoroacetate, NWTPQAMLYLKGAQ-NH2 trifluoroacetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C74H114N20O19S · xC2HF3O2
CAS Number:
Molecular Weight:
1619.88 (free base basis)
Assay:
≥95% (HPLC)
Form:
(Powder or Lyophilized powder or film)
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

(Powder or Lyophilized powder or film)

storage condition

desiccated

color

white to off-white

storage temp.

-10 to -25°C

General description

Spexin is a 4-amino acidpeptide, produced by neurons in the hypothalamus.

Application

Spexin for its potential roles in regulating appetite and metabolism,influencing reproductive hormones, in metabolic and endocrine research.

Biochem/physiol Actions

Spexin 1 (SPX) is a 14-aa amidated peptide that acts as an endogenousgalanin receptor GAL2/3 agonist (EC50 = 161 nM/GAL2, 626 nM/GAL3, >10µM/GAL1 by reporter assays using respective HEK293 transfectants) that inhibits facilitated fatty acid (FA) uptake in diet-induced obesity (DIO) mouse adipocytes cultures (0.1-1 ng/mL) and induces rat stomach muscle contraction ex vivo (EC50 = 750 nM). SPX reduces caloric intake in rats (∼32%; 35 µg/kg/day s.c.) with corresponding weight loss and significantly reduces the RER at night as well as increases locomotion in mice in vivo (25 µg/kg/day i.p.). Note: peptide sequence is conserved in human, murine, rat and many other mammalian species.

Disclaimer

Hygroscopic Store with desiccant


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library