Skip to Content
MilliporeSigma

SML4262

BDW568

≥98% (HPLC), powder

Synonym(s):

[(8,9-Dimethylthieno[3,2-e]-1,2,4-triazolo[4,3-c]pyrimidin-3-yl)thio]acetic acid methyl ester, Methyl 2-((8,9-dimethylthieno[3,2-e][1,2,4]triazolo[4,3-c]pyrimidin-3-yl)thio)acetate Acetic acid, 2-[(8,9-dimethylthieno[3,2-e]-1,2,4-triazolo[4,3-c]pyrimidin-3-yl)thio]-, methyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H12N4O2S2
CAS Number:
Molecular Weight:
308.38
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated, protect from light

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

General description

Ester precursor of the STING(A230/I230)-selective agonist BDW568-OH with no activity toward STING(G230).
BDW568 is an ester precursor of the STING(A230/I230)-selective agonist BDW568-OH with no activity toward STING(G230). Upon entering cells, BDW568 is hydrolyzed by carboxylesterase 1 (CES1) to the active metabolite BDW568-OH, which in turn induces cellular type-I interferon (IFN-I) signaling (EC50 = 7.6 μM by ISRE reporter assay using THP-1 cells, 4.1 μM using STING-/- THP-1 expressing STING with H71, A230, Q293).

Disclaimer

Hygroscopic Store with dessicant


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library