Skip to Content
MilliporeSigma

SML4263

NA-014

≥98% (HPLC)

Synonym(s):

2-(4-Cyanophenyl)-N-(4-fluorophenethyl)-2-(4-methylpiperazin-1-yl)acetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H25FN4O
CAS Number:
Molecular Weight:
380.46
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

NA-014 is a brain-penetrant piperazinyl-acetamide that functions as a highly selective positive allosteric modulator (PAM) of the excitatory amino acid transporter 2 (EAAT2), with no activity on EAAT1 and EAAT3. NA-014 effectively enhances EAAT2-mediated glutamate uptake, demonstrating an EC50 of 3.5 nM and an efficacy of 167.3% in EAAT1−3 transfected COS-7 cells, and an EC50 of 13.4 nM with 189.4% efficacy in glial cultures. NA-014 does not exhibit off-target effects on a broad panel of related receptors, including the hERG channel, displays good metabolic stability, possesses moderate water solubility, and shows favorable pharmacokinetic properties. Additionally, both enantiomers of NA-014 are active, binding at the allosteric site of human EAAT2 and demonstrating equivalent activity in functional assays designed to assess stimulation of EAAT2.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library