Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C42H38BF2N3OP·PF6
Molecular Weight:
825.52
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Quality Segment
assay
≥98% (HPLC)
form
powder
storage condition
desiccated
color
purple to black
solubility
DMSO: 2 mg/mL, clear
storage temp.
-10 to -25°C
Biochem/physiol Actions
MitoPerOx is a mitochondria-targeted lipid peroxidation (LPO) fluorescent ratiometric probe composed of the lipophilic triphenylphosphonium (TP) cation-conjugated C11-BODIPY(581/591) whose emission max shifts from ∼590 to ∼520 nm in response to LPO. Rapid mitochondria MitoPerOx uptake (within 1.5 min) facilitates the detection of mitochondrial lipid peroxidation in live cells by fluorimetry, confocal microscopy and epifluorescence imaging. Cells or isolated mitochondria are typically incubated with MitoPerOx (0.1-1 µM) for 0.5-1.5 hrs in the dark at 37 °C prior to washing and fluorescence detection (Ex/Em 580/600 nm & 490/520 nm; or by dual 520/590 nm detection with Ex 490 nm).
Disclaimer
Hygroscopic Store with desiccant
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.