Skip to Content
MilliporeSigma

SML4274

IDR-1002 trifluroacetate

95% (HPLC)

Synonym(s):

L-Valyl-L-glutaminyl-L-arginyl-L-tryptophyl-L-leucyl-L-isoleucyl-L-valyl-L-tryptophyl-L-arginyl-L-isoleucyl-L-arginyl-L-lysinamide trifluroacetate, VQRWLIVWRIRK-NH2 trifluroacetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C79H130N26O13 · xC2HF3O2
CAS Number:
Molecular Weight:
1652.04 (free base basis)
Assay:
95% (HPLC)
Form:
(Powder or Lyophilized powder or film)
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

95% (HPLC)

form

(Powder or Lyophilized powder or film)

storage condition

desiccated

color

white to off-white

storage temp.

-10 to -25°C

Biochem/physiol Actions

IDR-1002 is a synthetic analog of host defense peptides that exhibits broad-spectrum anti-infective properties, including immunomodulatory, anti-biofilm and antimicrobial activities. IDR-1002 potently enhances chemokine induction in vitro.

Disclaimer

Hygroscopic Store with desiccant


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library