Skip to Content
MilliporeSigma

SML4275

ACE-Okra Glow trifluoroacetate

≥98% (HPLC)

Synonym(s):

ACE trifluoroacetate, ACE-Okra FluoroForge trifluoroacetate, N-(2-(2-Aminoethoxy)ethyl)-2-cyano-3-(2,3,6,7-tetrahydro-1H,5H-pyrido[3,2,1-ij]quinolin-9-yl)acrylamide trifluoroacetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H26N4O2 · xC2HF3O2
Molecular Weight:
354.45 (free base basis)
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

-10 to -25°C

Biochem/physiol Actions

ACE (ACE-Okra Glow), a non-fluorescent molecular rotor, serves as the fluorogenic ligand for the Okra RNA aptamer (more than 400-fold fluorescence enhancement), forming the Okra505 complex that exhibits exceptional brightness and photostability in live-cell imaging. The Okra-ACE system demonstrates rapid binding kinetics (KD ≈ 1.2 nM; kon = 0.97 × 106 M−1 s−1 & koff = 1.0 × 10-3 s−1), low overall background fluorescence, pH resistant and good cell permeability, enabling robust visualization of mRNA dynamics, including detection of low-abundance mRNAs and observation of mRNA recruitment to subcellular structures like stress granules. Compatible with standard GFP filter sets, Okra505 allows for dual-color super-resolution imaging when combined with other fluorescent RNA aptamers, facilitating detailed studies of mRNA behavior and interactions in living cells while accounting for potential lysosomal localization. Excitation ~ 468 nm, emission ~ 505 nm & Φ = 0.93.

Disclaimer

Hygroscopic Store with dessicant


Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library