Skip to Content
MilliporeSigma

SML4280

ZT-1a

≥98% (HPLC)

Synonym(s):

5-chloro-N-(5-chloro-4-((4-chlorophenyl)(cyano)methyl)-2-methylphenyl)-2-hydroxybenzamide, SPAK Inhibitor ZT-1a

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H15Cl3N2O2
CAS Number:
Molecular Weight:
445.73
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

ZT-1a is a potent and selective, non-ATP competitive inhibitor of SPAK-NKCC1 signaling. It inhibits SPAK activity with an IC50 of 44.3µM. Blocking the SPAK–NKCC1 cascade prevents excessive ion influx (Na⁺, K⁺, Cl⁻), which is triggered by oxidative stress. Inhibition of this pathway helps maintain cellular ion homeostasis, reduces oxidative stress-induced apoptosis, and protects the integrity of the blood–CSF barrier. Blocking SPAK–NKCC1 also mitigates neuroinflammation by reducing immune cell infiltration, thus offering neuroprotection in both in vitro and in vivo models.


Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Codes

Hazard Classifications

Aquatic Acute 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library