Skip to Content
MilliporeSigma

SML4281

Ynamide 2e

≥97% (NMR), oil

Synonym(s):

N-Ethynyl-1,1,1-trifluoro-N-(2-hydroxyethyl)methanesulfonamidel

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H6F3NO3S
CAS Number:
Molecular Weight:
217.17
UNSPSC Code:
12352203
Assay:
≥97% (NMR)
Form:
oil
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (NMR)

form

oil

color

colorless to yellow

storage temp.

-10 to -25°C

Application

Ynamide 2e is suitable for use in the modification of Cys-containing proteins and antibodies.

Biochem/physiol Actions

Ynamide-2e, featuring a N-trifyl group, provides a highly efficient and chemoselective method for cysteine-specific modification in peptides and proteins. Ynamide-2e exclusively forms Z-isomer conjugates with superior stability under various conditions. This bioconjugation tool exhibits excellent thiol specificity, with a second-order rate constant of 0.2399 M−1 s−1 for glutathione, outperforming traditional reagents like iodoacetamide in reducing off-target labeling. Functioning efficiently in aqueous buffer, Ynamide-2e is compatible with a wide range of proteins, including antibodies, without compromising protein integrity or function. Its applicability in bottom-up proteomics workflows and potential for redox proteomics make it valuable for studying protein modifications and interactions.


Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Codes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library