Skip to Content
MilliporeSigma

SML4282

Clickable Antibody Rebridger, BUPY

≥98% (HPLC), powder, rebridging agent

Synonym(s):

3,5-Dibromo-1-(but-3-ynyl)pyrazin-1-ium triflate, 3,5-Dibromo-1-(but-3-yn-1-yl)pyrazin-1-ium trifluoromethanesulfonate, BUPY

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C8H7Br2N2·CF3SO3
Molecular Weight:
440.03
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

General description

BUPY, a novel dibromopyrazinium-based rebridging agent, offers rapid and efficient site-selective modification of antibody interchain disulfide bonds, achieving highly homogeneous conjugates with a degree of labeling of 4 and 97% rebridging efficiency. BUPY’s excellent aqueous stability, fast reactivity, and ability to function in slightly acidic conditions make it advantageous for antibody conjugation. The terminal alkyne handle enables facile click-chemistry functionalization with therapeutic or diagnostic payloads, while preserving the antibody′s target recognition capabilities. BUPY′s versatility and effectiveness position it as a valuable tool for researchers developing next-generation antibody-drug conjugates and diagnostic antibody probes.

Application

Clickable Antibody Rebridger, BUPY may be used to site-selectively reconnect reduced interchain cysteines on IgG antibodies, such as trastuzumab, enabling the creation of homogeneous conjugates that can be further modified with fluorophores or cytotoxic payloads. It may also be used in in the dSEC-MS workflow to quantify the efficiency and quality of rebridged antibodies, enabling real-time monitoring of antibody rebridging, assessment of critical quality attributes, and verification of full rebridging versus half-antibody species in BUPY-modified monoclonal antibodies.

Features and Benefits

  • Quick site-selective antibody modification.
  • Degree of labeling of 4.
  • 97% rebridging efficiency.
  • Excellent stability in water.
  • Quick reaction times.
  • Functions in slightly acidic conditions.
  • Terminal alkyne for click chemistry.
  • Maintains antibody target capabilities.
  • Ideal for antibody-drug conjugates.

Disclaimer

Hygroscopic Store with desiccant


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library