Skip to Content
MilliporeSigma

SML4284

Fumarate hydratase-IN-1

≥98% (HPLC), powder, fumarate hydratase inhibitor

Synonym(s):

rel- Ethyl (3S,3aR)-1-([1,1′-biphenyl]-4-ylmethyl)-3-(2-(methylamino)-2-oxoethyl)-2-oxo-1,2,3,4,5,6-hexahydro-3aH-indole-3a-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C27H30N2O4
CAS Number:
Molecular Weight:
446.54
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Application

Fumarate hydratase-IN-1 may be used:

  • to study its effects on mitochondrial bioenergetics and macrophage metabolism
  • to assess its regulatory effects in LPS-induced acute lung injury (ALI) models
  • to study the role of fumarate in immunomodulation and regulating type I interferon production after viral infection

Biochem/physiol Actions

FHIN1 exhibits nutrient-dependent antiproliferation activity in cancer cultures (mean IC50 = 2.2 µM in glucose-free L-15 medium; SW620, ACHN, HCT-116, PC3, SK-MEL-28) by targeting tricarboxylic acid (TCA) cycle without blocking mitochondrial electron-transport chain (ETC) or ATP synthase.


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library