Skip to Content
MilliporeSigma

SML4286

N-Acetyltaurine

≥95% (HPLC), powder, Taurine derivative

Synonym(s):

2-(Acetylamino)ethanesulfonic acid, [2-Acetylamino]ethanesulfonic acid, N-Acetyl-taurine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C4H9NO4S
CAS Number:
Molecular Weight:
167.18
MDL number:
Assay:
≥95% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

Application

N-Acetyltaurine (NacT) may be used as a marker for ethanol intake using the HILIC-based liquid chromatography methods and mass spectrometry.

Biochem/physiol Actions

N-Acetyltaurine (NAT) is a taurine-derived metabolite regulated by PTER (N-acetyltaurine hydrolase), which catalyzes its hydrolysis to taurine and acetate. NAT can be produced through multiple pathways, including the reverse reaction of PTER, catalyzing taurine acetylation under high acetate conditions, non-enzymatic condensation of taurine with acetyl-CoA, and gut microbiome-mediated production, suggesting microbiota involvement in its regulation. N-Acetyltaurine suppresses appetite through the GFRAL receptor, mitigates inflammation, preserves blood–CSF barrier integrity, and reduces oxidative stress. Acetyltaurine also reduces neuroinflammation by limiting immune cell infiltration and lowering pro-inflammatory cells like neutrophils and macrophages. In vivo studies show it reduces food intake, body weight, and adiposity, while improving glucose tolerance, supporting overall metabolic health.


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library