Skip to Content
MilliporeSigma

SML4290

AU-24118 hydrochloride

≥98% (HPLC)

Synonym(s):

3-(4-(1′-(4-(3-Amino-6-(2-hydroxyphenyl)pyridazin-4-yl)phenyl)-[1,4′-bipiperidin]-4-yl)phenoxy)piperidine-2,6-dione hydrochloride, AU 24118 hydrochloride, AU24118 hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C37H40N6O4 · xHCl
CAS Number:
Molecular Weight:
632.75 (free base basis)
UNSPSC Code:
41116107
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

light yellow to orange

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

AU-24118 is an orally active, potent and selective SWI/SNF ATPase degrader composed of an E3 ligase cereblon (CRBN) ligand linked to the SMARCA2/4 and PBRM1 (PB1)-selective bromodomain (BrD) inhibitor SGC-SMARCA-BRDVIII. AU-24118 induces near complete degradation of cellular SMARCA2/4 and PBRM1 (4-8h, 100 nM, 22Rv1 cells) and blocks SWI/SNF-dependent survival in cancer cultures (IC50 <100 nM post 5-day treatment; C42B, LNCaP, 22Rv1, VCaP). AU-24118 induces tumor regression in a castration-resistant prostate cancer (CRPC) VCaP xenograft model (15 mg/kg 3x/wk via p.o.) and is enhanced with concurrent AR antagonist enzalutamide treatment (10 mg/kg 5x/wk via p.o.).

Disclaimer

Hygroscopic, store with desiccant


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library