Skip to Content
MilliporeSigma

SML4294

SBI-115

≥98% (HPLC), powder, TGR5/GPBAR1 antagonist

Synonym(s):

3-Methylphenyl 5-chloro-2-(ethylsulfonyl)-4-pyrimidinecarboxylate, m-tolyl 5-chloro-2-(ethylsulfonyl) pyrimidine-4-carboxylate, SBI-115, TGR5 Antagonist, SBI115, SBI 115

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H13ClN2O4S
CAS Number:
Molecular Weight:
340.78
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Application

SBI-115 may be used in cell proliferation and cyclic adenosine monophosphate (cAMP) assay to determine its effects on rodents with polycystic liver diseases and, to explore the mechanisms of TGR5 in inhibiting NLR family pyrin domain containing 3 (NLRP3) inflammasome under inflammatory conditions.

Biochem/physiol Actions

SBI-115, a specific TGR5/GPBAR1 antagonist, blocks agonist activation, inhibiting TGR5-Gαs-cAMP signaling and reducing intracellular cAMP in cystic cholangiocytes. This antagonism dose-dependently inhibits agonist-induced proliferation and reduces 3D cholangiocyte spheroid growth in vitro, enabling SBI-115 to attenuate Polycystic Liver Disease (PLD) pathology by modulating cAMP. Beyond PLD, SBI-115′s TGR5 modulation impacts inflammation by offsetting bile acid suppression of IL-1β/NLRP3 in macrophages and influences macrophage M2 polarization in non-small cell lung cancer, promoting antitumor immunity in vitro. Additionally, SBI-115 (1 µM) elevates triglycerides and reduces P-PPARA in fatty-degenerated hepatocytes, implicating TGR5 in lipid metabolism.


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library