Skip to Content
MilliporeSigma

SML4295

TAC

≥98% (HPLC)

Synonym(s):

N-(3-Chloro-4-fluorophenyl)-4-fluoro-3,5-dimethylbenzenesulfonamide, TERT activator compound

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H12ClF2NO2S
CAS Number:
Molecular Weight:
331.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

General description

Cell penetrant, selective and potent activator of telomerase reverse transcriptase (TERT).
TAC (TERT activator compound) is a cell penetrant, selective and potent activator of telomerase reverse transcriptase (TERT) that upregulates TERT transcription through activation of the MEK/ERK/AP-1 cascade. It induces TERT expression in WS fibroblasts (0.5 µM) and increases telomere synthesis in primary human cells. TAC alleviates neuroinflammation, increases neurotrophic factors, stimulates adult neurogenesis, and preserves cognitive function without toxicity in brain of naturally aged mice.


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library