Skip to Content
MilliporeSigma

SML4298

STING degrader AK59

≥98% (HPLC)

Synonym(s):

AK59, AK59-51TB, N-(5-(pyridin-3-yl)quinolin-8-yl)-4-(trifluoromethyl)benzenesulfonamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H14F3N3O2S
Molecular Weight:
429.41
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

STING degrader AK59 is a potent inhibitor of the cGAS/STING pathway (IC50 = 5.19 µM) by induction of STING interaction with ligase HERC4 (a HECT-domain E3 ligase) followed by degradation of STING. AK59 facilitates HERC4 to transfer ubiquitin directly to Lysine 150 of STING, which initiates the degradation. It is effective against wild-type STING and STING-associated vasculopathy (SAVI) mutant STING proteins in human CD14+ monocytic cells.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library