Skip to Content
MilliporeSigma

SML4301

Desthiobiotin-NHS

≥95% (HPLC), powder, amine-reactive NHS tag

Synonym(s):

2,5-Dioxopyrrolidin-1-yl 6-((4R,5S)-5-methyl-2-oxoimidazolidin-4-yl)hexanoate, Desthiobiotin N-hydroxysuccinimidyl ester, NHS-Desthiobiotin,N-Hydroxysuccinimido dethiobiotinate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H21N3O5
CAS Number:
Molecular Weight:
311.33
Assay:
≥95% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Application

Desthiobiotin-NHS may be used in protein modification, bioconjugation, and pull-down assays.

Biochem/physiol Actions

Desthiobiotin-NHS, an amine-reactive NHS tag, facilitates efficient and reversible protein enrichment. The desthiobiotin handle, a structural analog of biotin lacking a sulfur atom, exhibits a reduced binding affinity for avidin/streptavidin (µM KD) compared to biotin (pM KD), enabling gentle elution through competitive displacement with biotin or by weakening the interaction with mild alkali, thereby preserving protein integrity. This NHS tag readily conjugates to protein amines, enabling flexible, multi-cycle binding, washing, and elution. Note: the lower affinity of the desthiobiotin handle requires careful optimization to minimize non-specific interactions and potential loss during washes compared to biotin-streptavidin systems.


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library