Skip to Content
MilliporeSigma

SML4302

PR-104

≥95% (HPLC)

Synonym(s):

2-[N-(2-Bromoethyl)-2,4-dinitro-6-[[[2-(phosphonooxy)ethyl]amino]carbonyl]anilino]ethyl methanesulfonate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H20BrN4O12PS
CAS Number:
Molecular Weight:
579.27
UNSPSC Code:
12352200
Assay:
≥95% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

PR-104 is an anticancer agent activated by hypoxia, a condition common in human solid tumors. It has een used for research in multiple tumor xenograft models. PR-104 is converted to the lipophilic dintrobenzamide mustards alcohol PR-104A.

Disclaimer

Hygroscopic, store with desiccant


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library