Skip to Content
MilliporeSigma

SML4306

4-Azido-L-phenylalanine hydrochloride

≥98% (HPLC)

Synonym(s):

(S)-2-Amino-3-(4-azidophenyl)propanoic acid hydrochloride, H-Phe(4-N3)-OH HCl, 4-Azidophenylalanine HCl, p-Azido-L-phenylalanine HCl, pAzF

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H10N4O2·HCl
CAS Number:
Molecular Weight:
242.66
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

4-Azido-L-phenylalanine (pAzF), an unnatural amino acid featuring a bioorthogonal azide handle, is site-specifically incorporated into proteins via genetic code expansion, providing a unique modification site. This incorporated azide enables precise attachment of probes (dyes, tags, therapeutics) through bioorthogonal click chemistry or photo-crosslinking, yielding proteins with novel functionalities for imaging, interaction studies, and creating biomaterials/bioconjugates. Consequently, 4-Azido-L-phenylalanine (pAzF) is a powerful tool in chemical biology and biotechnology for protein engineering, functional analysis, and therapeutic applications across diverse cellular systems.

Disclaimer

Hygroscopic, Store with desiccant


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library