Skip to Content
MilliporeSigma

SML4307

Lipoamide

≥98% (HPLC), powder, coenzyme

Synonym(s):

1,2-Dithiolane-3-valeramide, (±)-1,2-Dithiolane-3-pentanamide, (±)-1,2-Dithiolane-3-valeramide, (±)-Thioctic acid amide, DL-α-Lipoic acid amide, DL-6,8-Thioctamide, Ticolin; Vitamin N, α-Lipoamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C8H15NOS2
CAS Number:
Molecular Weight:
205.34
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Application

Lipoamide may be used:


  • as a small‑molecule antioxidant / neuroprotective agent (a neutral amide derivative of lipoic acid) to evaluate protection against oxidative‑stress‑induced neuronal cell damage
  • as a stimulator of mitochondrial biogenesis in differentiated 3T3‑L1 adipocytes to explore potential benefits for metabolic dysfunction
  • to study its effects on hypertensive myocardial hypertrophy using rat models

Biochem/physiol Actions

Lipoamide, a lipoic acid acting as a coenzyme for of enzymes involved in amino acid metabolism, is a well-tolerated modulator of stress granule condensation that specifically prevents cytoplasmic condensation of stress granule proteins. Lipoamide stabilizes intrinsically disordered domain-containing proteins, including SRSF1 and SFPQ, and reverses stress granule formation in vitro. Lipoamide ameliorates aging-associated aggregation of a stress granulereporter protein, improves neuronal morphology and recovers motor defects caused by amyotrophic lateral sclerosis (ALS)-associated FUS and TDP-43 mutants in fly (Drosophila melanogaster) models of ALS.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library