Skip to Content
MilliporeSigma

SML4309

Sulfo-NHS-LC-Biotin

≥95% (HPLC)

Synonym(s):

Sulfo-NHS-LC-Biotin sodium salt, Sulfosuccinimidyl 6-(biotinamido) hexanoate, sodium salt, 1-[[6-[[5-[(3aS,4S,6aR)-Hexahydro-2-oxo-1H-thieno[3,4-d]imidazol-4-yl]-1-oxopentyl]amino]-1-oxohexyl]oxy]-2,5-dioxo-3-pyrrolidinesulfonic acid, monosodium salt, 2,5-Dioxo-1-((6-(5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamido)hexanoyl)oxy)pyrrolidine-3-sulfonate, sodium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H29NaN4O9S2
CAS Number:
Molecular Weight:
556.59
UNSPSC Code:
12352200
Assay:
≥95% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Sulfo-NHS-LC-biotin is a membrane-permeant derivatization agent utilized for covalent labeling of primary amines via amide linkage formation through its NHS ester. Sulfo-NHS-LC-biotin allows enrichment of derivatized proteins through streptavidin-coated beads or surface for downstream applications, such as target proteins identification and quantification.

Disclaimer

Hygroscopic Store with desiccant


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library