Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C6H12N4O2 · xHCl
Molecular Weight:
172.19 (free base basis)
MDL number:
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Quality Segment
assay
≥98% (HPLC)
form
powder
storage condition
desiccated
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
-10 to -25°C
Biochem/physiol Actions
L-Azidonorleucine (ANL) is a noncanonical, slow to metabolize, and clickable amino acid that can be incorporated to proteins in cells that express a mutant methionyl-tRNA synthetase (MetRS). The newly synthesized MetRS-expressing cell-derived proteins from cell mixtures can then be enriched and examined in a cell-type-specific manner. The azido handle in ANL can be selectively conjugated to dyes and functionalized beads to enable visualization of proteins by FUNCAT (Fluorescence non-canonical amino acid tagging), and to purify proteins using BONCAT (bio-orthogonal non-canonical amino acid tagging).
Disclaimer
Hygroscopic Store with desiccant
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.